4-(Aminomethyl)pyridin-2-amine 95

Supplier: Matrix Scientific

073343-250MG 073343-1G 073343-5G
102529-664EA 355.38 USD
102529-664 102529-666 102529-668
4-(Aminomethyl)pyridin-2-amine 95
4-(Aminomethyl)pyridin-2-amine

Hazard: Irritant

Formula: C₆H₉N₃
MW: 123.1558 g/mol
MDL Number: MFCD06213870
CAS Number: 199296-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR