4-Bromo-1,2-dihydropyridazine-3,6-dione ≥95%

Supplier: Matrix Scientific

073342-1G 073342-5G
102529-660EA 175.38 USD
102529-660 102529-662
4-Bromo-1,2-dihydropyridazine-3,6-dione ≥95%
4-Bromo-1,2-dihydropyridazine-3,6-dione

Hazard: Irritant

Formula: C₄H₃BrN₂O₂
MDL Number: MFCD12923098
CAS Number: 15456-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR