7-Bromochroman-4-one ≥95%
Supplier: Matrix Scientific
073339-1G
073339-5G
102529-644EA
67.69
USD
102529-644
102529-646
7-Bromochroman-4-one ≥95%
7-Bromochroman-4-one
Hazard: Irritant
Formula:
C₉H₇BrO₂ |
MDL Number:
MFCD09744055 CAS Number: 18442-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...