2-Acetyl-5-bromopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(5-Bromopyridin-2-yl)ethanone

073338-1G 073338-5G
102529-638EA 75.38 USD
102529-638 102529-640
2-Acetyl-5-bromopyridine ≥95%
2-Acetyl-5-bromopyridine

Hazard: Irritant/Light Sensitive/Stored under Argon

Formula: C₇H₆BrNO
MW: 200.0300 g/mol
MDL Number: MFCD04974523
CAS Number: 214701-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR