N-Methyl-5-bromo-4,7-diazaindole ≥95%

Supplier: Matrix Scientific

073335-250MG 073335-1G
102529-622EA 684.62 USD
102529-622 102529-624
N-Methyl-5-bromo-4,7-diazaindole ≥95%
N-Methyl-5-bromo-4,7-diazaindole

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
MDL Number: MFCD12964054
CAS Number: 1217090-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR