2-Bromo-5-tosyl-5H-pyrrolo[2,3-b]pyrazine ≥95%
Supplier: Matrix Scientific
073334-250MG
073334-1G
073334-5G
102529-616EA
456.92
USD
102529-616
102529-618
102529-620
2-Bromo-5-tosyl-5H-pyrrolo[2,3-b]pyrazine ≥95%
2-Bromo-5-tosyl-5H-pyrrolo[2,3-b]pyrazine
Hazard: Irritant
Formula:
C₁₃H₁₀BrN₃O₂S MW: 352.2100 g/mol |
MDL Number:
MFCD12964053 CAS Number: 1201186-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...