tert-Butyl (pyrrolidin-2-ylmethyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Boc-aminomethyl-pyrrolidine

073333-1G 073333-5G
102529-612EA 92.31 USD
102529-612 102529-614
tert-Butyl (pyrrolidin-2-ylmethyl)carbamate ≥95%
tert-Butyl (pyrrolidin-2-ylmethyl)carbamate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
MDL Number: MFCD06658352
CAS Number: 149649-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR