(R)-(-)-1-Methoxy-2-propanol ≥95%
Supplier: Matrix Scientific
073330-5G
073330-10G
073330-100G
102529-596EA
30.77
USD
102529-596
102529-598
102529-600
(R)-(-)-1-Methoxy-2-propanol ≥95%
(R)-(-)-1-Methoxy-2-propanol
Hazard: Flammable
Formula:
C₄H₁₀O₂ Boiling Pt: 119-121°C Density: 0.921 Flash Pt: 33°C |
MDL Number:
MFCD01632587 CAS Number: 4984-22-9 UN: 3092 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...