4-Methoxyphthalic acid ≥95%
Supplier: Matrix Scientific
073327-1G
073327-5G
073327-25G
102529-580EA
76.92
USD
102529-580
102529-582
102833-622
4-Methoxyphthalic acid ≥95%
4-Methoxyphthalic acid
Hazard: Irritant
Formula:
C₉H₈O₅ |
MDL Number:
MFCD00094739 CAS Number: 1885-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...