6-Chloro-2-hydroxymethylpyridine hydrochloride ≥95%

Supplier: Matrix Scientific

073323-1G 073323-5G
102529-560EA 304.62 USD
102529-560 102529-562
6-Chloro-2-hydroxymethylpyridine hydrochloride ≥95%
6-Chloro-2-hydroxymethylpyridine hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂NO
MW: 180.0300 g/mol
MDL Number: MFCD08690070
CAS Number: 83782-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR