tert-Butyl (2-(methylamino)ethyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-N'-methyl-1,2-diaminoethane, tert-Butyl N-(2-Methylaminoethyl)carbamate, N-(2-Methylaminoethyl)carbamic acid tert-Butyl ester, N-Boc-N'-methylethylenediamine, N-(tert-Butoxycarbonyl)-N'-methylethylenediamine, N-(tert-Butoxycarbonyl)-N'-methyl-1,2-diaminoethane

073322-1G 073322-5G
102529-556EA 30.77 USD
102529-556 102529-558
tert-Butyl (2-(methylamino)ethyl)carbamate ≥95%
tert-Butyl (2-(methylamino)ethyl)carbamate

Hazard: Irritant

Formula: C₈H₁₈N₂O₂
MW: 174.2407 g/mol
MDL Number: MFCD06808585
CAS Number: 122734-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR