(S)-N-Boc-3-methylmorpholine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-N-Boc-3-Methylmorpholine

073321-250MG 073321-1G
102529-552EA 1223.08 USD
102529-552 102529-554
(S)-N-Boc-3-methylmorpholine ≥95%
(S)-N-Boc-3-methylmorpholine

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.2600 g/mol
MDL Number: MFCD12964052
CAS Number: 1022094-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR