(R)-N-Boc-3-methylmorpholine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-N-Boc-3-Methylmorpholine

073320-250MG 073320-1G
102529-548EA 1223.08 USD
102529-548 102529-550
(R)-N-Boc-3-methylmorpholine ≥95%
(R)-N-Boc-3-methylmorpholine

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD16038029
CAS Number: 1022093-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR