5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

073319-1G 073319-5G 073319-10G
102529-542EA 107.69 USD
102529-542 102529-544 102529-546
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid ≥95%
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₁Cl₃N₂O₂
MW: 381.6400 g/mol
MDL Number: MFCD06411080
CAS Number: 162758-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR