3-Bromo-5-(4-fluorophenyl)pyridine ≥95%

Supplier: Matrix Scientific

073318-250MG 073318-1G 073318-5G
102529-536EA 498.46 USD
102529-536 102529-538 102529-540
3-Bromo-5-(4-fluorophenyl)pyridine ≥95%
3-Bromo-5-(4-fluorophenyl)pyridine

Hazard: Irritant

Formula: C11H7BrFN
MW: 252.08 g/mol
MDL Number: MFCD09800589
CAS Number: 675590-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR