4-Chloroquinoline ≥95%

Supplier: Matrix Scientific

073317-5G 073317-25G
102529-532EA 64.62 USD
102529-532 102833-620
4-Chloroquinoline ≥95%
4-Chloroquinoline

Hazard: Irritant

Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 260-261°C
Melting Pt: 28-31°C
Density: 1.25
MDL Number: MFCD00006773
CAS Number: 611-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR