Ethyl-5-bromopyrimidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-bromopyrimidine-2-carboxylate

073315-1G 073315-5G 073315-10G
102529-520EA 533.85 USD
102529-520 102529-522 102529-524
Ethyl-5-bromopyrimidine-2-carboxylate ≥95%
Ethyl-5-bromopyrimidine-2-carboxylate

Hazard: Irritant

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
MDL Number: MFCD12964050
CAS Number: 1197193-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR