Methyl-5-bromopyrimidine-2-carboxyate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-Bromopyrimidine-2-carboxylate, 5-Bromopyrimidine-2-carboxylic acid methyll ester

073314-1G 073314-5G 073314-10G
102529-514EA 533.85 USD
102529-514 102529-516 102529-518
Methyl-5-bromopyrimidine-2-carboxyate ≥95%
Methyl-5-bromopyrimidine-2-carboxyate

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD11111606
CAS Number: 89581-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR