5-Amino-4-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073312-250MG 073312-1G 073312-5G
102529-502EA 427.69 USD
102529-502 102529-504 102529-506
5-Amino-4-methylpyrimidine ≥95%
5-Amino-4-methylpyrimidine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.1300 g/mol
MDL Number: MFCD09038780
CAS Number: 3438-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR