5-Methylnicotinaldehyde ≥95%

Supplier: Matrix Scientific

073311-250MG 073311-1G
102529-496EA 158.46 USD
102529-496 102529-498
5-Methylnicotinaldehyde ≥95%
5-Methylnicotinaldehyde

Hazard: Irritant

Formula: C7H7NO
MDL Number: MFCD08272078
CAS Number: 100910-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR