4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073310-1G 073310-5G
102529-492EA 75.38 USD
102529-492 102529-494
4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%
4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₇H₆ClN₃
MW: 167.6000 g/mol
MDL Number: MFCD09746275
CAS Number: 71149-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR