5-Chloropyrazine-2-carboxylic acid hydrazide ≥95%

Supplier: Matrix Scientific

073308-250MG 073308-1G 073308-5G
102529-482EA 640 USD
102529-482 102529-484 102529-486
5-Chloropyrazine-2-carboxylic acid hydrazide ≥95%
5-Chloropyrazine-2-carboxylic acid hydrazide

Hazard: Irritant

Formula: C5H5ClN4O
MDL Number: MFCD12964049
CAS Number: 848952-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR