tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate ≥95%

Supplier: Matrix Scientific

073302-250MG 073302-1G 073302-5G
102529-452EA 350.77 USD
102529-452 102529-454 102529-456
tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate ≥95%
tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate

Hazard: Irritant

Formula: C₉H₁₃BrN₂O₂S
MDL Number: MFCD09878588
CAS Number: 1000576-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR