3-Acetamido-4-methyl-2-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

073300-1G 073300-5G 073300-10G
102529-440EA 355.38 USD
102529-440 102529-442 102529-444
3-Acetamido-4-methyl-2-nitrobenzoic acid ≥95%
3-Acetamido-4-methyl-2-nitrobenzoic acid

Hazard: Irritant

Formula: C10H10N2O5
MDL Number: MFCD00755098
CAS Number: 7356-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR