4-Methyl-1H-benzimidazole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
073299-250MG
073299-1G
073299-5G
102529-434EA
400
USD
102529-434
102529-436
102529-438
4-Methyl-1H-benzimidazole-2-carboxylic acid ≥95%
4-Methyl-1H-benzimidazole-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₈N₂O₂ MW: 176.1720 g/mol |
MDL Number:
MFCD09751735 CAS Number: 673487-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...