2-Chloro-5-methylpyrazine ≥95%
Supplier: Matrix Scientific
073298-250MG
073298-1G
102529-428EA
92.31
USD
102529-428
102529-430
2-Chloro-5-methylpyrazine ≥95%
2-Chloro-5-methylpyrazine
Hazard: Irritant
Formula:
C₅H₅ClN₂ MW: 128.5600 g/mol |
MDL Number:
MFCD00055039 CAS Number: 59303-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...