tert-Butyl furan-3-ylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Aminofuran, N-BOC protected

073296-250MG 073296-1G 073296-5G
102529-416EA 427.69 USD
102529-416 102529-418 102529-420
tert-Butyl furan-3-ylcarbamate ≥95%
tert-Butyl furan-3-ylcarbamate

Hazard: Irritant

Formula: C₉H₁₃NO₃
MW: 183.2044 g/mol
MDL Number: MFCD09839085
CAS Number: 56267-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR