5-Bromobenzofuran-3(2H)-one ≥95%

Supplier: Matrix Scientific

073295-250MG 073295-1G 073295-5G
102529-410EA 355.38 USD
102529-410 102529-412 102529-414
5-Bromobenzofuran-3(2H)-one ≥95%
5-Bromobenzofuran-3(2H)-one

Hazard: Irritant

Formula: C₈H₅BrO₂
MDL Number: MFCD06738724
CAS Number: 54450-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR