3-Amino-2,6-difluorobenzoic acid ≥95%

Supplier: Matrix Scientific

073294-1G 073294-5G
102529-406EA 75.38 USD
102529-406 102529-408
3-Amino-2,6-difluorobenzoic acid ≥95%
3-Amino-2,6-difluorobenzoic acid

Hazard: Irritant

Formula: C7H5F2NO2
MDL Number: MFCD07368833
CAS Number: 83141-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR