5-Methylpyrazin-2(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-5-methylpyrazine

073293-1G 073293-5G
102529-402EA 423.08 USD
102529-402 102529-404
5-Methylpyrazin-2(1H)-one ≥95%
5-Methylpyrazin-2(1H)-one

Hazard: Irritant

Formula: C5H6N2O
MW: 110.1139 g/mol
MDL Number: MFCD01693937
CAS Number: 20721-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR