2-Fluoro-3-nitrophenol ≥95%

Supplier: Matrix Scientific

073292-250MG 073292-1G 073292-5G
102529-396EA 184.62 USD
102529-396 102529-398 102529-400
2-Fluoro-3-nitrophenol ≥95%
2-Fluoro-3-nitrophenol

Hazard: Irritant

Formula: C₆H₄FNO₃
MW: 157.1000 g/mol
MDL Number: MFCD11042443
CAS Number: 179816-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR