2-Amino-5-bromobenzo[d]thiazole-3-oxide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromobenzo[d]thiazole 3-oxide

073291-1G 073291-5G
102529-392EA 307.69 USD
102529-392 102529-394
2-Amino-5-bromobenzo[d]thiazole-3-oxide ≥95%
2-Amino-5-bromobenzo[d]thiazole-3-oxide

Hazard: Irritant

Formula: C₇H₅BrN₂OS
MW: 245.1 g/mol
MDL Number: MFCD12406142
CAS Number: 1216671-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR