2-Chloro-7-iodo-5H-pyrrolo[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073290-250MG 073290-1G 073290-5G
102529-386EA 1110.77 USD
102529-386 102529-388 102529-390
2-Chloro-7-iodo-5H-pyrrolo[3,2-d]pyrimidine ≥95%
2-Chloro-7-iodo-5H-pyrrolo[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₃ClIN₃
CAS Number: 1152475-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR