2-Chloro-7-iodothieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073289-250MG 073289-1G
102529-382EA 684.62 USD
102529-382 102529-384
2-Chloro-7-iodothieno[3,2-d]pyrimidine ≥95%
2-Chloro-7-iodothieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₂ClIN₂S
MDL Number: MFCD12406129
CAS Number: 1152475-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR