7-Bromo-1H-imidazo[4,5-c]pyridine ≥95%

Supplier: Matrix Scientific

073288-250MG 073288-1G 073288-5G
102529-376EA 640 USD
102529-376 102529-378 102529-380
7-Bromo-1H-imidazo[4,5-c]pyridine ≥95%
7-Bromo-1H-imidazo[4,5-c]pyridine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Melting Pt: 292…294 °C
MDL Number: MFCD09834585
CAS Number: 90993-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR