5-Bromo-2-chloro-4-methyl-3-nitropyridine ≥95%

Supplier: Matrix Scientific

073287-1G 073287-5G
102529-372EA 498.46 USD
102529-372 102529-374
5-Bromo-2-chloro-4-methyl-3-nitropyridine ≥95%
5-Bromo-2-chloro-4-methyl-3-nitropyridine

Hazard: Irritant

Formula: C₆H₄BrClN₂O₂
MW: 251.47 g/mol
MDL Number: MFCD06659505
CAS Number: 884495-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR