5-Bromopyridine-3,4-diamine ≥95%

Supplier: Matrix Scientific

073285-1G 073285-5G
102529-364EA 98.46 USD
102529-364 102529-366
5-Bromopyridine-3,4-diamine ≥95%
5-Bromopyridine-3,4-diamine

Hazard: Irritant

Formula: C₅H₆BrN₃
MW: 188.03 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD01692479
CAS Number: 4635-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR