2,4-Dichloro-6-morpholino-1,3,5-triazine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4,6-Dichloro-1,3,5-triazin-2-yl)morpholine, 2,4-Dichloro-6-(4-morpholinyl)-1,3,5-triazine

073282-1G 073282-5G
102529-348EA 90.77 USD
102529-348 102529-350
2,4-Dichloro-6-morpholino-1,3,5-triazine ≥95%
2,4-Dichloro-6-morpholino-1,3,5-triazine

Hazard: Irritant

Formula: C₇H₈Cl₂N₄O
MW: 235.0706 g/mol
MDL Number: MFCD00460178
CAS Number: 6601-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR