5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

073281-250MG 073281-1G 073281-5G
102529-342EA 498.46 USD
102529-342 102529-344 102529-346
5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid ≥95%
5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₇N₃O₂S
MW: 185.2000 g/mol
MDL Number: MFCD12407810
CAS Number: 100130-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR