Methyl-2-amino-5-methoxybenzoate ≥95%

Supplier: Matrix Scientific

073280-1G 073280-5G
102529-338EA 207.69 USD
102529-338 102529-340
Methyl-2-amino-5-methoxybenzoate ≥95%
Methyl-2-amino-5-methoxybenzoate

Hazard: Irritant

Formula: C₉H₁₁NO₃
MDL Number: MFCD09038482
CAS Number: 2475-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR