Methyl-2-amino-5-hydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxyanthranilic acid methyll ester, Methyl 5-Hydroxyanthranilate, Methyl 2-amino-5-hydroxybenzoate, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic acid methyll ester

073279-1G 073279-5G
102529-332EA 710.77 USD
102529-332 102529-334
Methyl-2-amino-5-hydroxybenzoate ≥95%
Methyl-2-amino-5-hydroxybenzoate

Hazard: Irritant

Formula: C₈H₉NO₃
MDL Number: MFCD05863263
CAS Number: 1882-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR