5-Chloropyrimidin-4-ol ≥95%

Supplier: Matrix Scientific

073276-1G 073276-5G
102529-316EA 275.38 USD
102529-316 102529-318
5-Chloropyrimidin-4-ol ≥95%
5-Chloropyrimidin-4-ol

Hazard: Irritant

Formula: C4H3ClN2O
CAS Number: 4349-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR