Methyl-5-bromo-1H-indole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-1H-indole-3-carboxylate

073274-1G 073274-5G
102529-306EA 126.15 USD
102529-306 102529-308
Methyl-5-bromo-1H-indole-3-carboxylate ≥95%
Methyl-5-bromo-1H-indole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
MDL Number: MFCD06203728
CAS Number: 773873-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR