2-(7-(Dimethylamino)-2-oxo-2H-chromen-4-yl)acetic acid ≥95%

Supplier: Matrix Scientific

073273-250MG 073273-1G
102529-302EA 924.62 USD
102529-302 102529-304
2-(7-(Dimethylamino)-2-oxo-2H-chromen-4-yl)acetic acid ≥95%
2-(7-(Dimethylamino)-2-oxo-2H-chromen-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₄
MW: 247.2466 g/mol
MDL Number: MFCD00857826
CAS Number: 80883-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR