7-Quinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

073270-1G 073270-5G
102529-286EA 303.08 USD
102529-286 102529-288
7-Quinolinecarboxylic acid ≥95%
7-Quinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂
MW: 173.1700 g/mol
MDL Number: MFCD00047617
CAS Number: 1078-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR