2-Bromo-6-chlorobenzo[d]thiazole ≥95%

Supplier: Matrix Scientific

073268-1G 073268-5G
102529-278EA 284.62 USD
102529-278 102529-280
2-Bromo-6-chlorobenzo[d]thiazole ≥95%
2-Bromo-6-chlorobenzo[d]thiazole

Hazard: Irritant

Formula: C₇H₃BrClNS
MW: 248.5300 g/mol
MDL Number: MFCD07783781
CAS Number: 3507-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR