2-Nitro-4-pyridinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Nitroisonicotinic acid

073267-1G 073267-5G
102529-274EA 150.77 USD
102529-274 102529-276
2-Nitro-4-pyridinecarboxylic acid ≥95%
2-Nitro-4-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
MDL Number: MFCD04114160
CAS Number: 33225-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR