4-Bromo-2-(methylsulfonyl)pyrimidine ≥95%

Supplier: Matrix Scientific

073266-250MG 073266-1G
102529-268EA 630.77 USD
102529-268 102529-270
4-Bromo-2-(methylsulfonyl)pyrimidine ≥95%
4-Bromo-2-(methylsulfonyl)pyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂O₂S
MW: 237.0744 g/mol
MDL Number: MFCD12406118
CAS Number: 1208538-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR