2-Chloro-6-methoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

073265-1G 073265-5G 073265-10G
102529-262EA 60 USD
102529-262 102529-264 102529-266
2-Chloro-6-methoxybenzaldehyde ≥95%
2-Chloro-6-methoxybenzaldehyde

Hazard: Irritant

Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
MDL Number: MFCD08059233
CAS Number: 29866-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR