5-Chloropyrazine-2-carboxamide ≥95%

Supplier: Matrix Scientific

073264-250MG 073264-1G 073264-5G
102529-256EA 400 USD
102529-256 102529-258 102529-260
5-Chloropyrazine-2-carboxamide ≥95%
5-Chloropyrazine-2-carboxamide

Hazard: Irritant

Formula: C5H4ClN3O
MW: 157.5600 g/mol
MDL Number: MFCD10686873
CAS Number: 21279-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR